Learn More
TCR alpha/beta Antibody (BMA031) [Alexa Fluor™ 405], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-43550AF405
Product Code. 30280986
  • 855.00 € / 0.10mL
Description
TCR alpha/beta Monoclonal antibody specifically detects TCR alpha/beta in Human samples. It is validated for Immunoprecipitation,Neutralization,Serum,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TCR alpha/beta
Monoclonal
Alexa Fluor 405
C), D, FLJ22602, J, MGC117436, MGC22624, MGC23964, MGC71411, T cell receptor alpha locus, T-cell antigen receptor, alpha polypeptide, T-cell receptor, alpha (V, TCRA, TCRD, TRA
The original antibody was generated by immunizing mice with human perihperal blood T-lymphocytes.
0.1 mL
Adaptive Immunity, Cell Biology, Diabetes Research, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases
6955
Store at 4⪚C in the dark.
IgG
Immunoprecipitation, Neutralization, Immunocytochemistry/Immunofluorescence
BMA031
50mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
Human
Purified