missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR alpha/beta Antibody (Desire-1) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
262.00 € - 618.00 €
Specifications
| Antigen | TCR alpha/beta |
|---|---|
| Clone | Desire-1 |
| Concentration | 1 mg/mL |
| Applications | Flow Cytometry, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18622179
|
Novus Biologicals
NBP3-09020-0.025mg |
0.025 mg |
262.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18646598
|
Novus Biologicals
NBP3-09020-0.2mg |
0.2 mg |
618.00 €
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TCR alpha/beta Monoclonal antibody specifically detects TCR alpha/beta in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
| TCR alpha/beta | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| C), D, FLJ22602, J, MGC117436, MGC22624, MGC23964, MGC71411, T cell receptor alpha locus, T-cell antigen receptor, alpha polypeptide, T-cell receptor, alpha (V, TCRA, TCRD, TRA | |
| mAb desire-1 was obtained after fusion of spleen cells from a BALB.K mouse immunised twice with KB5-C20 clone cells in RPMI 1640 medium, with non-Ig-producing myeloma cells X63-Ag8.653. Desire-1 was selected for its capacity to inhibit selectively the H-2Kb-specific cytolysis by clone KB5-C20. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Desire-1 | |
| Flow Cytometry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Cell Biology, Diabetes Research, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| PBS | |
| 6955 | |
| IgG1 κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title