missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR beta Armenian Hamster, APC, Clone: H57-597, enQuireBio™
Armenian Hamster Monoclonal Antibody
Brand: EnquireBio QAB79-APC-100ug
This item is not returnable.
View return policy
Specifications
| TCR beta | |
| Monoclonal | |
| 0.5mg/mL | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
| Armenian Hamster | |
| 100 μg | |
| Primary | |
| Store at 2-4°C. |
| Flow Cytometry | |
| H57-597 | |
| APC | |
| Tcrb | |
| Protein A/G Purified | |
| RUO | |
| 21577 | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction