missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TCR V alpha 2 Monoclonal Antibody (B20.1), NovaFluor™ Red 700, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
178.00 € - 344.00 €
Specifications
| Antigen | TCR V alpha 2 |
|---|---|
| Clone | B20.1 |
| Concentration | 0.1 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30055648
|
Invitrogen™
M033T03R03-A |
100 μg |
344.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30055110
|
Invitrogen™
M033T02R03-A |
25 μg |
178.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TCR V alpha 2 Monoclonal antibody specifically detects TCR V alpha 2 in Mouse samples. It is validated for Flow CytometrySpecifications
| TCR V alpha 2 | |
| 0.1 mg/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| TCRVA; TCRVA2; V alpha 2; V alpha-2 TCR; Va 2; Va2; Valpha 2; Valpha2 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| B20.1 | |
| Flow Cytometry | |
| NovaFluor Red 700 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| IgG2a λ | |
| Affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title