missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TCR V alpha 2 Monoclonal Antibody (B20.1), NovaFluor™ Yellow 660, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

178.00 € - 344.00 €

Specifications

Antigen TCR V alpha 2
Clone B20.1
Concentration 0.1 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30056205
View Documents
Invitrogen™
M033T03Y04-A
100 μg
344.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30055498
View Documents
Invitrogen™
M033T02Y04-A
25 μg
178.00 €
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TCR V alpha 2 Monoclonal antibody specifically detects TCR V alpha 2 in Mouse samples. It is validated for Flow Cytometry
Specifications

Specifications

TCR V alpha 2
0.1 mg/mL
Monoclonal
Rat
Mouse
TCRVA; TCRVA2; V alpha 2; V alpha-2 TCR; Va 2; Va2; Valpha 2; Valpha2
Primary
4°C, store in dark, DO NOT FREEZE!
B20.1
Flow Cytometry
NovaFluor Yellow 660
RUO
PBS with 0.09% sodium azide; pH 7.2
IgG2a λ
Affinity chromatography
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.