missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR V beta 3 Antibody (JOVI.3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
214.00 € - 382.00 €
Specifications
| Antigen | TCR V beta 3 |
|---|---|
| Clone | JOVI.3 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Functional, CyTOF-ready |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Functional Assay, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18686526
|
Novus Biologicals
NBP2-50356-0.025ml |
0.025 mL |
214.00 €
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18642829
|
Novus Biologicals
NBP2-50356 |
0.1 mL |
382.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TCR V beta 3 Monoclonal antibody specifically detects TCR V beta 3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Functional Assay, CyTOFSpecifications
| TCR V beta 3 | |
| 1 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Functional Assay, CyTOF | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Cell Biology, Diabetes Research, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| PBS | |
| T cell receptor beta locus, T-cell antigen receptor, beta polypeptide, T-cell receptor, beta cluster, T-cell receptor, beta cluster, TCRB, TRB | |
| Thymus, spleen and mesenteric lymph nodes isolated from a mouse transgenic for human Vb3 TcR. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| JOVI.3 | |
| Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Functional, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P09693 | |
| 6957 | |
| IgG2a | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title