missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TDO2 Antibody (998604) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB9768P
This item is not returnable.
View return policy
Description
TDO2 Monoclonal antibody specifically detects TDO2 in Human samples. It is validated for Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| TDO2 | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF | |
| 998604 | |
| Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| TDOTO, TPH2EC 1.13.11.11, TRPOtryptophanase, Tryptamin 2,3-dioxygenase, tryptophan 2,3-dioxygenase, Tryptophan oxygenase, Tryptophan pyrrolase, Tryptophanase | |
| E. coli-derived human TDO2, Leu18-Phe388, Accession # P48775 | |
| 0.1 mL | |
| Immunology, Lipid and Metabolism, Neuroscience | |
| 6999 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction