Learn More
TDP-43 Rabbit anti-Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10782-2-AP-20UL
 View more versions of this product
Product Code. 16857393
  • 145.00 € / 20µL
Description
Proteintech's 10782-2-AP antibody is a rabbit polyclonal antibody recognizing N-terminal TDP-43. It recognizes the intact 45 kDa protein as well as all post translationally modified and truncated forms in multiple applications. Various forms of TDP-
TDP43 was first identified as a novel cellular protein that binds to HIV-1 virus TAR DNA sequence motifs and acts a transcriptional repressor to the HIV-1 LTR. Later experiments revealed that TDP43 also regulates the splicing of exon 9 of the cystic fibrosis transmembrane conductance regular (CFTR), most likely through the association with the UG repeats at the 3"e; splice site. A hyperphosphorylated, ubiquitinated, and cleaved form of TDP43 known as pathologic TDP43 is the major disease protein in ubiquitin-positive, tau-, and alpha-synuclein-negative frontotemporal dementia (FLTD-U). TDP43 is not related to TRBP1, and RNA binding protein that binds HIV-1 TAR RNA sequences.
Specifications
Specifications
TDP-43
Polyclonal
Unconjugated
TARDBP
ALS10, TAR DNA binding protein, TAR DNA binding protein 43, TARDBP, TDP 43, TDP43
Rabbit
Antigen Affinity Chromatography
RUO
230908, 23435, 298648, 325052
-20°C
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry, Flow Cytometry
0.31 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q13148, Q921F2
tardbp
TDP-43 Fusion Protein Ag1231
20 μL
Primary
Rat, Zebrafish, Mouse, Human