missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TDRG1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41197
This item is not returnable.
View return policy
Description
TDRG1 Polyclonal specifically detects TDRG1 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| TDRG1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| TDRG1 | |
| Antibody was raised against a 17 amino acid peptide near the center of human TDRG1. The immunogen is located within amino acids 40 - 90 of TDRG1. Amino Acid Squence: CIKISSTPPSQSRLDPQI | |
| Affinity Purified | |
| RUO | |
| 732253 | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA | |
| LINC00532, Long Intergenic Non-Protein Coding RNA 532, Testis Development Related 1 (Non-Protein Coding) | |
| Rabbit | |
| 11 kDa | |
| 0.1 mg | |
| Primary | |
| TDRG1 antibody is human reactive. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction