missing translation for 'onlineSavingsMsg'
Learn More

TDZD-8, MedChemExpress

Product Code. 16465266
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16465266 100 mg 100mg
16464358 10 mg 10mg
16420687 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16465266 Supplier Medchem Express Supplier No. HY11012
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress TDZD-8 is an inhibitor of GSK-3β, with an IC50 of 2 μM; TDZD-8 shows less potent activities against Cdk-1/cyclin B, CK-II, PKA, and PKC, with all IC50s of >100 μM.

TDZD-8 is an inhibitor of GSK-3β, with an IC50 of 2 μM

Specifications

Chemical Name or Material TDZD-8
Physical Form Solid
CAS 327036-89-5
Color Light Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C10H10N2O2S
For Use With (Application) Cancer-Kinase/protease
Synonym GSK-3β Inhibitor I NP 01139
Solubility Information DMSO : ≥ 100 mg/mL (449.92 mM)
SMILES O=C(N1CC2=CC=CC=C2)N(C)SC1=O
Molecular Weight (g/mol) 222.26
Formula Weight 222.26
Percent Purity 99.54%
Grade Research
Show More Show Less

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.