missing translation for 'onlineSavingsMsg'
Learn More

TE Buffer, 1X Solution, Molecular Biology Grade, Ultrapure

Product Code. 15875228 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mL
10 x 1 mL
500 mL
Packaging:
Nalgene bottle
Unit Size:
100mL
500mL
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15875228 10 x 1 mL Nalgene bottle Pack of 10
15661509 100 mL Nalgene bottle 100mL
15875418 500 mL Nalgene bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15875228 Supplier Thermo Scientific Supplier No. J75893.X^
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Use directly to resuspend and/or dilute purified DNA or RNA | CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol

Use directly to resuspend and/or dilute purified DNA or RNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 1185-53-1
Molecular Formula C4H12ClNO3
Molecular Weight (g/mol) 157.594
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
PubChem CID 93573
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
SMILES C(C(CO)(CO)N)O.Cl

Specifica

Physical Form Solution
Quantity 10 x 1 mL
Packaging Nalgene bottle
Grade Molecular Biology
Chemical Name or Material TE Buffer

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.