missing translation for 'onlineSavingsMsg'
Learn More

TE Buffer, 1X Solution pH 8.0, Low EDTA, Molecular Biology Grade

Product Code. 15684018 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Nalgene bottle
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15684018 500 mL Nalgene bottle 500mL
15746272 100 mL Nalgene bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15684018 Supplier Thermo Scientific Supplier No. J75793.AP
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use directly to re-suspend and/or dilute purified DNA or RNA | CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol

Use directly to re-suspend and/or dilute purified DNA or RNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185-53-1
Molecular Formula C4H12ClNO3
Molecular Weight (g/mol) 157.594
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
PubChem CID 93573
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
SMILES C(C(CO)(CO)N)O.Cl

Specifications

Concentration 1X
pH 8
Physical Form Solution
Quantity 500 mL
Packaging Nalgene bottle
Grade Molecular Biology
Chemical Name or Material TE Buffer

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.