missing translation for 'onlineSavingsMsg'
Learn More

Melford TE Buffer 20X Solution pH 8.0

Product Code. 30018848
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
This item is not returnable. View return policy

Product Code. 30018848

Brand: Melford T32200250.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

Formulation:

TRIS HCI: 200 mM

EDTA: 20 mM

Specifications

CAS 77-86-1, 1185-53-1, 6381-92-6
Quantity 250 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.