missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford TE Buffer 20X Solution pH 8.0
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
Formulation:
TRIS HCI: 200 mM
EDTA: 20 mM
Specifications
Specifications
| CAS | 77-86-1, 1185-53-1, 6381-92-6 |
| Quantity | 250 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction