missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tebuconazole

Product Code. 16600842 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16600842 5 g 5g
16690832 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16600842 Supplier Thermo Scientific Chemicals Supplier No. 463260050
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 107534-96-3 | C16H22ClN3O | 307.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107534-96-3
Molecular Formula C16H22ClN3O
Molecular Weight (g/mol) 307.82
MDL Number MFCD02674797
InChI Key PXMNMQRDXWABCY-UHFFFAOYNA-N
IUPAC Name 1-(4-chlorophenyl)-4,4-dimethyl-3-[(1H-1,2,4-triazol-1-yl)methyl]pentan-3-ol
SMILES CC(C)(C)C(O)(CCC1=CC=C(Cl)C=C1)CN1C=NC=N1

Specifications

Melting Point 102.0°C to 105.0°C
Flash Point >100°C
Quantity 5 g
Formula Weight 307.82
Chemical Name or Material Tebuconazole
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tebuconazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H361d-Suspected of damaging the unborn child.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Tebuconazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.