missing translation for 'onlineSavingsMsg'
Learn More

Tebuconazole, MedChemExpress

Product Code. 16451446
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
1 g
Unit Size:
1g
200mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16451446 200 mg 200mg
16442289 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16451446 Supplier Medchem Express Supplier No. HYB0852
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Tebuconazole is an agricultural azole fungicide which can also inhibit CYP51 with IC50s of 0.9 and 1.3 μM for Candida albicans CYP51 (CaCYP51) and truncated Homo sapiens CYP51 (δ60HsCYP51), respectively.

Tebuconazole is an agricultural azole fungicide which can also inhibit CYP51 with IC50s of 0.9 and 1.3 μM for Candida albicans CYP51 (CaCYP51) and truncated Homo sapiens CYP51 (δ60HsCYP51), respectively.

Specifications

Chemical Name or Material Tebuconazole
Physical Form Solid
CAS 107534-96-3
Color White
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C16H22ClN3O
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 50 mg/mL (162.43 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CC(C)(C)C(O)(CCC1=CC=C(Cl)C=C1)CN2C=NC=N2
Molecular Weight (g/mol) 307.82
Formula Weight 307.82
Percent Purity 99.64%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.