missing translation for 'onlineSavingsMsg'
Learn More

Tebuconazole, TRC

Product Code. 30135508
Change view
Click to view available options
Quantity:
25 mg
1 g
10 g
Unit Size:
10g
1g
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135508 25 mg 25mg
30132779 10 g 10g
17965115 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135508 Supplier Toronto Research Chemicals Supplier No. TRCT01300025MG
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107534-96-3
Molecular Formula C16H22ClN3O
Molecular Weight (g/mol) 307.82
Synonym 1H-1,2,4-Triazole-1-ethanol, α-[2-(4-chlorophenyl)ethyl]-α-(1,1-dimethylethyl)- (9CI, ACI), 1H-1,2,4-Triazole-1-ethanol, α-[2-(4-chlorophenyl)ethyl]-α-(1,1-dimethylethyl)-, (±)- (ZCI), α-[2-(4-Chlorophenyl)ethyl]-α-(1,1-dimethylethyl)-1H-1,2,4-triazole-1-ethanol (ACI), (±)-Tebuconazole, 1-(4-Chlorophenyl)-4,4-dimethyl-3-(1,2,4-triazole-1-yl-methyl)pentane-3-ol, Atlas, BAY-HWG 1608, Corail, Dedalus 25 WDG, Elite, Elite (fungicide), Ethyltrianol, Etiltrianol, Fenetrazole, Folicur, Folicur 200CE, Folicur 200EC, Folicur 3.6F, Gelseal, Gelseal (fungicide), Greenseal, Hao Li-ke, Helicur 250EW, Horizon, Horizon (fungicide), Horizont, HWG 1608, Microban S 2142, Microban TZ, Miresa, Monsoon, ONLY ONE Flowable, Orius, Orius (fungicide), Orius 25EC, Preventol A 8, Raxil, Raxil 2DS, Raxil Ultra, Rival 200EC, Riza, Rosacur, S 2142, Silvacure, Sparta, Sparta 250EW, Tebu 250EW, Tebu 60, Tebuconazole, Terbutrazole, Timbertreat T
IUPAC Name 1-(4-chlorophenyl)-4,4-dimethyl-3-(1,2,4-triazol-1-ylmethyl)pentan-3-ol
SMILES CC(C)(C)C(O)(CCc1ccc(Cl)cc1)Cn2cncn2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C16H22ClN3O/c1-15(2,3)16(21,10-20-12-18-11-19-20)9-8-13-4-6-14(17)7-5-13/h4-7,11-12,21H,8-10H2,1-3H3
Formula Weight 307.1451
Chemical Name or Material Tebuconazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.