missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tebufloquin metabolite M1, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Tebufloquin Metabolite M1 |
| CAS | 1403836-05-4 |
| Analyte or Component Names | Tebufloquin Metabolite M1 |
| Shipping Condition | +20°C |
| Molecular Formula | C15 H18 F N O |
| Certifications/Compliance | ISO/IEC 17025 |
| Synonym | 6-(1,1-Dimethylethyl)-8-fluoro-2,3-dimethyl-4(1H)-quinolinone, Tebufloquin Metabolite M1; |
| InChI Formula | InChI=1S/C15H18FNO/c1-8-9(2)17-13-11(14(8)18)6-10(7-12(13)16)15(3,4)5/h6-7H,1-5H3,(H,17,18) |
| SMILES | CC1=C(C)C(=O)c2cc(cc(F)c2N1)C(C)(C)C |
| IUPAC Name | 6-tert-butyl-8-fluoro-2,3-dimethyl-1H-quinolin-4-one |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?