missing translation for 'onlineSavingsMsg'
Learn More

Technical Buffer pH 11.00, Mettler Toledo™

Product Code. 11962535
Change view
Click to view available options
Quantity:
6 x 250 mL
Unit Size:
1500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11962535 6 x 250 mL 1500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11962535 Supplier Mettler Toledo Online Supplier No. 51350026
  • ONLINE EXCLUSIVE PRICE
    105.00 €
    92.72 € / 1500mL
    Save 12.28 €
    11% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Traceable to the primary standards of reference materials selected by NIST.

  • pH 11.0
TRUSTED_SUSTAINABILITY

Specifications

Description Technical buffer pH 11.00, 6 x 250mL
pH 11
Quantity 6 x 250 mL
Solution Type Buffer
Packaging 6x 250 mL
compliance-icons
Product Identifier
  • Mettler-Toledo technical buffer, pH 11.00, 6x250 mL
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: diisopropylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.