missing translation for 'onlineSavingsMsg'
Learn More

Technical Buffer for pH DKD Certified pH 7.00, METTLER TOLEDO™

Product Code. 10711955
Change view
Click to view available options
Quantity:
250 mL
Packaging:
1Each
6x 250 mL
Unit Size:
1500mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10711955 250 mL 6x 250 mL 1500mL
10709883 250 mL 1Each 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10711955 Supplier Mettler Toledo Online Supplier No. 51350044
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The DKD (Deutscher Kalibrierdienst, German Calibration Service) certified buffers are the perfect product for regulated industries.

  • pH 7.00
TRUSTED_SUSTAINABILITY

Specifications

Description Certified buffer pH 7.00, 6 x 250mL
pH 7.00
Quantity 250 mL
Packaging 6x 250 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.