missing translation for 'onlineSavingsMsg'
Learn More

Technical Buffer Solution pH 7, WTW™
GREENER_CHOICE

Product Code. 11745164
Change view
Click to view available options
Quantity:
250 mL
1000 mL
Packaging:
1 Bottle
10 Bottles
25 Bottles
Unit Size:
250mL
Each
Pack of 25
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11745164 250 mL 1 Bottle 250mL
10216304 250 mL 25 Bottles Pack of 25
10595505 1000 mL 10 Bottles Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11745164 Supplier WTW Supplier No. 108802
From Chemicals to Chromatography
Use promo code 27991
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Precise and traceable to NIST (National Institute of Standards and Technology), their accuracy is ± 0,03 pH.

  • Precise and traceable to NIST (National Institute of Standards and Technology, USA in bulk size of 250 ml with built-in dosing vessel standard buffer
  • Life up to 3 years (unopened)
  • Thrifty use
  • Easiest handling by means of dosing device
TRUSTED_SUSTAINABILITY

Specifications

pH 7
Physical Form Liquid
Quantity 250 mL
Packaging 1 Bottle
Traceability to NIST Yes
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Zinc acetate dihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash skin thoroughly after handling.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P391-Collect spillage.
Supplemental information
  • MIXTURE LIST-Contain: Zinc di(acetate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.