missing translation for 'onlineSavingsMsg'
Learn More

Technical pH Buffer Solutions, METTLER TOLEDO™

Product Code. 10598281
Change view
Click to view available options
Color:
Blue
Colorless
Green
Red
pH:
2.00
4.01
7.00
9.21
10.00
11.00
Unit Size:
250mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color pH Unit Size
10598281 Colorless 2.00 250mL
10383191 Blue 9.21 250mL
10343291 Colorless 10.00 250mL
10634091 Colorless 11.00 250mL
10683701 Green 7.00 250mL
10201841 Red 4.01 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10598281 Supplier Mettler Toledo Online Supplier No. 51350002
Spotlight Offers
Use promo code 28038
Se alle kampagner
 Restricted Item
In Stock
Læg i indkøbskurv
Edge
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

Provides a reliable all-around pH buffer. Mettler Toledo™ Technical pH Buffer Solution are specified to two decimal places and are traceable to NIST standards. Color-coded solution comes in a handy 250mL plastic bottle with no-fuss labels.

pH measurements are only as accurate as the buffer solutions used for calibration purposes. These technical-grade buffer solutions provide a reliable pH buffer for regular use with most applications. (pH electrodes should be calibrated at least once a day prior to measurements and after every cleaning, regeneration or long-term storage.)

  • NIST traceable and specified to two decimal places
  • Handy screw-top plastic bottles for easy handling
  • Includes a test certificate and no-fuss label with a temperature table
  • SDS and label contains information according to GHS (Globally Harmonized System)
TRUSTED_SUSTAINABILITY

Tekniske data

Color Colorless
For Use With (Application) pH measurement, electrode calibration
For Use With (Equipment) pH Electrode
pH 2.00
Quantity 250 mL
Odor Odorless
Packaging Plastic, screw-cap bottle
Traceability to NIST Yes
Grade Technical
Concentration or Composition (by Analyte or Components) 0.5–1% Citric Acid, 0.01–0.1% Hydrogen chloride
compliance-icons
Produktidentifikator
  • Technical buffer pH 2.00
Signalord
  • Warning
Farekategori
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Faresætning
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Sikkerhedssætning
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplerende oplysninger
  • MIXTURE LIST-Contain: Citric acid anhydrous, Hydrogen chlorideanhydrous
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.