missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TECPR1 Polyclonal Antibody

Description
Peptide sequence: YTKDKKWNSC VRRRKWIRYR RYKSRDIWAK IPSKDDPKEL PDPFNDLSVG Sequence homology: Dog: 93%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.
This gene encodes a tethering factor involved in autophagy. The encoded protein is found at autolysosomes, and is involved in targeting protein aggregates, damaged mitochondria, and bacterial pathogens for autophagy.
Specifications
Specifications
| Antigen | TECPR1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | TECPR1 |
| Gene Accession No. | Q7Z6L1 |
| Gene Alias | 2210010N04Rik; KIAA1358; LOW QUALITY PROTEIN: tectonin beta-propeller repeat-containing protein 1; mKIAA1358; RGD1306873; TECPR1; tectonin beta-propeller repeat containing 1; tectonin beta-propeller repeat-containing protein 1 |
| Gene Symbols | TECPR1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?