missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEF1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
214.00 € - 493.00 €
Specifications
| Antigen | TEF1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin, Chromatin Immunoprecipitation (ChIP) 1:20-1:50 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), ChIP Assay |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18607332
|
Novus Biologicals
NBP2-94035-0.02ml |
0.02 mL |
214.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18669741
|
Novus Biologicals
NBP2-94035-0.1ml |
0.1 mL |
493.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TEF1 Polyclonal antibody specifically detects TEF1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), ChIP assay (ChIP)Specifications
| TEF1 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), ChIP Assay | |
| Unconjugated | |
| Rabbit | |
| Cardiovascular Biology, Cell Biology | |
| PBS (pH 7.3), 50% glycerol | |
| 7003 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin, Chromatin Immunoprecipitation (ChIP) 1:20-1:50 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| NTEF-1, protein GT-IIC, TEA domain family member 1 (SV40 transcriptional enhancer factor), TEAD-1 | |
| A synthetic peptide corresponding to a sequence within amino acids 200-300 of human TEF1 (NP_068780.2). APSVPAWQGRSIGTTKLRLVEFSAFLEQQRDPDSYNKHLFVHIGHANHSYSDPLLESVDIRQIYDKFPEKKGGLKELFGKGPQNAFFLVKFWADLNCNIQD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title