missing translation for 'onlineSavingsMsg'
Learn More

Tegobuvir, MedChemExpress

Product Code. 16490387
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16490387 50 mg 50mg
16456747 5 mg 5mg
16444038 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16490387 Supplier Medchem Express Supplier No. HY10544
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Tegobuvir is a specific, covalent inhibitor of the HCV NS5B polymerase.

Tegobuvir is a specific, covalent inhibitor of the HCV NS5B polymerase.

Specifications

Chemical Name or Material Tegobuvir
CAS 1000787-75-6
Color White
Purity Grade Notes Research
Molecular Formula C25H14F7N5
Quantity 50 mg
Synonym GS 333126 GS-9190
Solubility Information DMSO : ≥ 50 mg/mL (96.64 mM)
SMILES FC1=CC=CC=C1C2=NC3=CN(CC4=NN=C(C5=CC=C(C(F)(F)F)C=C5C(F)(F)F)C=C4)C=CC3=N2
Molecular Weight (g/mol) 517.4
Formula Weight 517.4
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.