missing translation for 'onlineSavingsMsg'
Learn More

Telaglenastat, MedChemExpress

Product Code. p-200098866
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491277 50 mg 50mg
16455786 100 mg 100mg
16435048 10 mg 10mg
16487767 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16491277 Supplier Medchem Express Supplier No. HY12248

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

MedChemExpress Telaglenastat (CB-839) is a first-in-class, selective, reversible and orally active glutaminase 1 (GLS1) inhibitor. Telaglenastat selectively inhibits GLS1 splice variants KGA (kidney-type glutaminase) and GAC (glutaminase C) compared to GLS2. The IC50s are 23 nM and 28 nM for endogenous glutaminase in mouse kidney and brain, respectively. Telaglenastat inudces autophagy and has antitumor activity.

Telaglenastat (CB-839) is a first-in-class, selective, reversible and orally active glutaminase 1 (GLS1) inhibitor. Telaglenastat selectively inhibits GLS1 splice variants KGA (kidney-type glutaminase) and GAC (glutaminase C) compared to GLS2. The IC50s are 23 nM and 28 nM for endogenous glutaminase in mouse kidney and brain, respectively. Telaglenastat inudces autophagy and has antitumor activity.

Spezifikation

Chemical Name or Material Telaglenastat
CAS 1439399-58-2
Color Light Brown
Purity Grade Notes Research
Molecular Formula C26H24F3N7O3S
Quantity 50 mg
Synonym CB-839
Solubility Information DMSO : 6.29 mg/mL (11.00 mM; Need ultrasonic)
SMILES O=C(NC1=NN=C(CCCCC2=NN=C(NC(CC3=CC=CC(OC(F)(F)F)=C3)=O)C=C2)S1)CC4=NC=CC=C4
Molecular Weight (g/mol) 571.57
Formula Weight 571.57
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Mehr anzeigen Weniger anzeigen

Research purposes only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.