missing translation for 'onlineSavingsMsg'
Learn More

Temocapril hydrochloride, MedChemExpress

Product Code. 16448468
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16448468 10 mg 10mg
16484347 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16448468 Supplier Medchem Express Supplier No. HYB0384
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Temocapril hydrochloride is an orally active angiotensin-converting enzyme (ACE) inhibitor. Temocapril hydrochloride can be used for the research of hypertension, congestive heart failure, acute myocardial infarction, insulin resistance, and renal diseases.

Temocapril hydrochloride is an orally active angiotensin-converting enzyme (ACE) inhibitor. Temocapril hydrochloride can be used for the research of hypertension, congestive heart failure, acute myocardial infarction, insulin resistance, and renal diseases.

Specifications

Chemical Name or Material Temocapril hydrochloride
Physical Form Powder
CAS 110221-44-8
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C23H29ClN2O5S2
For Use With (Application) COVID-19-anti-virus
Solubility Information DMSO : 62.5 mg/mL (121.82 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 2.27 mg/mL (4.42 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(O)CN1C[C@@H](C2=CC=CS2)SC[C@H](N[C@H](C(OCC)=O)CCC3=CC=CC=C3)C1=O.Cl
Molecular Weight (g/mol) 513.07
Formula Weight 513.07
Percent Purity 99.63%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.