missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tempol, MedChemExpress
Click to view available options
Quantity:
200 mg
1 g
Unit Size:
1g
200mg
Description
Tempol is a general superoxide dismutase (SOD)-mimetic drug that efficiently neutralizes reactive oxygen species (ROS).
Specifications
Specifications
| Chemical Name or Material | Tempol |
| Physical Form | Crystals |
| CAS | 2226-96-2 |
| Color | Orange |
| Quantity | 200 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C9H18NO2* |
| For Use With (Application) | Cancer-programmed cell death |
| Synonym | 4-Hydroxy-TEMPO |
| Solubility Information | H2O : 12.5 mg/mL (72.57 mM; ultrasonic and warming and heat to 60°C) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction