missing translation for 'onlineSavingsMsg'
Learn More

Temsirolimus, MedChemExpress

Product Code. 16418148
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16418148 10 mg 10mg
16483756 25 mg 25mg
16477896 100 mg 100mg
16460598 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16418148 Supplier Medchem Express Supplier No. HY50910

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Temsirolimus is an inhibitor of mTOR with an IC50 of 1.76 μM. Temsirolimus activates autophagy and prevents deterioration of cardiac function in animal model.

Temsirolimus is an inhibitor of mTOR with an IC50 of 1.76 μM. Temsirolimus activates autophagy and prevents deterioration of cardiac function in animal model.

Specifications

Chemical Name or Material Temsirolimus
Physical Form Powder
CAS 162635-04-3
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C56H87NO16
For Use With (Application) Cancer-Kinase/protease
Synonym CCI-779
Solubility Information DMSO : 250 mg/mL (242.65 mM; Need ultrasonic) ∣Ethanol : 200 mg/mL (194.12 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C([C@@]1(O)[C@@H](CC[C@@H](C[C@@H](/C(C)=C/C=C/C=C/[C@H](C[C@@H](C)C([C@@H]([C@@H](/C(C)=C/[C@H]2C)O)OC)=O)C)OC)O1)C)C(N3CCCC[C@H]3C(O[C@@H](CC2=O)[C@@H](C[C@H]4C[C@H]([C@H](OC(C(C)(CO)CO)=O)CC4)OC)C)=O)=O
Molecular Weight (g/mol) 1030.29
Formula Weight 1030.29
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.