missing translation for 'onlineSavingsMsg'
Learn More

Tenofovir Disoproxil fumarate, MedChemExpress

Product Code. 16476326
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16476326 100 mg 100mg
16471706 500 mg 500mg
16451106 200 mg 200mg
16442007 50 mg 50mg
16435858 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16476326 Supplier Medchem Express Supplier No. HY13782
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Tenofovir Disoproxil fumarate is a nucleotide reverse transcriptase inhibitor used to treat HIV and chronic Hepatitis B.

Tenofovir Disoproxil fumarate is a nucleotide reverse transcriptase inhibitor used to treat HIV and chronic Hepatitis B.

Specifications

Chemical Name or Material Tenofovir Disoproxil fumarate
Physical Form Solid
CAS 202138-50-9
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C23H34N5O14P
For Use With (Application) COVID-19-anti-virus
Synonym Tenofovir DFBis(POC)-PMPA fumarateGS 4331 fumarate
Solubility Information DMSO : ≥ 50 mg/mL (78.68 mM) ∣H2O : 16.67 mg/mL (26.23 mM; Need ultrasonic and warming)
SMILES O=C(OC(C)C)OCOP(OCOC(OC(C)C)=O)(CO[C@H](C)CN1C=NC2=C(N)N=CN=C12)=O.O=C(O)/C=C/C(O)=O
Molecular Weight (g/mol) 635.51
Formula Weight 635.51
Percent Purity 98.76%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.