missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti- Teprotumumab (Anti-Idiotype) Antibody, R&D Systems™

Mouse Monoclonal Antibody
201.00 € - 418.00 €
Specifications
| Antigen | Teprotumumab |
|---|---|
| Clone | 975547 |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, Binding Inhibition 2.5 ug/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18763809
![]() |
R&D Systems
MAB96341-SP |
25 μg |
201.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18777399
![]() |
R&D Systems
MAB96341-100 |
100 μg |
418.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Teprotumumab Monoclonal specifically detects Teprotumumab in Multi-Species samples. It is validated for Flow Cytometry, Binding Inhibition.Specifications
| Teprotumumab | |
| Flow Cytometry 0.25 ug/10^6 cells, Binding Inhibition 2.5 ug/mL | |
| Monoclonal | |
| Supernatant | |
| Multi-species | |
| Full-length Teprotumumab | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects Teprotumumab in direct ELISAs. |
| 975547 | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title