missing translation for 'onlineSavingsMsg'
Learn More

TER-119/Erythroid cells Rat anti-Mouse, BV421, Clone: TER-119, BD Horizon™

Rat Monoclonal Antibody

Brand:  BD Biosciences 566248

Product Code. 15814879

  • ONLINE EXCLUSIVE PRICE
    190.00 €
    181.90 € / Each
    Save 8.10 €
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TER-119/Erythroid cells Monoclonal antibody specifically detects TER-119/Erythroid cells in Mouse samples. It is validated for Flow cytometry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TER-119/Erythroid cells
Monoclonal
0.2mg/mL
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Rat
25 μg
Cell Biology
Mouse
Purified
Flow Cytometry
TER-119
Brilliant Violet 421
Lymphocyte antigen 76; Ly76; Ly-76; TER-119; Ter119
Mouse Fetal Liver
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions and unconjugated antibody and free BD Horizon™ BV421 were removed.
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Reseach Use Only.