Learn More
Isofield™ Tera 230 Validated Sterile Tie-On Cleanroom Facemask - Grade A/B Compatible
BIOPHARMA
Product Code. 30178873
Change view
Click to view available options
Size:
Wide
Unit Size:
Pack of 500
Product Code. Size Unit Size
30178873 Wide Pack of 500
1 options
Product Code. 30178873 Supplier Isofield™ Supplier No. 60131
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
Estimated Shipment: 18-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Extra wide validated sterile facemask. The Tera is made from gamma compatible materials ensuring low/no odor in use. The individually packed Tera has excellent bacterial and particle filtration efficiency and is GMP Grade A compatible.
  • Validated sterile tie-on cleanroom facemask
  • 230 mm length with malleable nose strip
  • 3-layer non-woven configuration
  • Highly efficient bacterial and particle filtration
  • Gamma irradiation, minimum 25 kGy
  • Sterility Assurance Level 10-6
  • Particle Shedding (Helmke Drum) Category I Individually packed pouches printed with IPA resistant ink to maintain traceability
Specifications
Product Type Cleanroom Face Mask
Color White
Fastening Type Ties
Material Polyethylene, Polypropylene
Size Wide
Sterility Sterile
Style Pleated