missing translation for 'onlineSavingsMsg'
Learn More

Terbinafine hydrochloride, MedChemExpress

Product Code. 16487276
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
Unit Size:
100mg
200mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16487276 100 mg 100mg
16401256 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16487276 Supplier Medchem Express Supplier No. HY17395100mg
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Terbinafine hydrochloride (TDT 067 hydrochloride) is an orally active and potent antifungal agent. Terbinafine hydrochloride is a potent non-competitive inhibitor of squalene epoxidase from Candida, with a Ki of 30 nM. Terbinafine hydrochloride also shows antibacterial activity against certain Gram-positive and Gram-negative bacteria.

Terbinafine hydrochloride (TDT 067 hydrochloride) is an orally active and potent antifungal agent. Terbinafine hydrochloride is a potent non-competitive inhibitor of squalene epoxidase from Candida, with a Ki of 30 nM. Terbinafine hydrochloride also shows antibacterial activity against certain Gram-positive and Gram-negative bacteria.

Specifications

Chemical Name or Material Terbinafine hydrochloride
Physical Form Solid
CAS 78628-80-5
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C21H26ClN
For Use With (Application) Neuroscience-Neuromodulation
Synonym TDT 067 hydrochloride
Solubility Information DMSO : 60 mg/mL (182.99 mM; Need ultrasonic) ∣H2O : 1 mg/mL (3.05 mM; ultrasonic and warming and heat to 60°C)
SMILES CN(C/C([H])=C([H])/C#CC(C)(C)C)CC1=CC=CC2=CC=CC=C21.[H]Cl
Molecular Weight (g/mol) 327.89
Formula Weight 327.89
Percent Purity 99.78%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.