missing translation for 'onlineSavingsMsg'
Learn More

Terbium Internal Standard: Tb @ 10 μg/mL in 2% HNO3, VHG™

Product Code. 17983546
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17983546 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17983546 Supplier VHG Labs Supplier No. VHGLISATB100
 Restricted Item
Estimated Shipment: 14-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Terbium Internal Standard: Tb @ 10 μg/mL in 2% HNO3
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Terbium
CAS 7440-27-9
Methods ICP-MS
Analyte or Component Names Terbium
Shipping Condition Room Temperature
Molecular Formula Tb
Certifications/Compliance ISO 17034
Synonym Terbium ingot, Terbium foil
InChI Formula InChI=1S/Tb
SMILES [Tb]
IUPAC Name terbium
Solvent or Matrix Dilute HNO3
Molecular Weight (g/mol) 158.925
Quantity 100 mL
Formula Weight 158.925 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.