missing translation for 'onlineSavingsMsg'
Learn More

Terbium solution for ICP/MS 100mg/L HNO3 matrix, Fisher Chemical™
BIOPHARMA

Product Code. 12977303 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12977303 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12977303 Supplier Fisher Chemical Supplier No. S 458 100
  • ONLINE EXCLUSIVE PRICE
    73.85 €
    57.55 € / 100mL
    Save 16.30 €
    22% Off
 Restricted Item
Estimated Shipment: 25-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Bottle
CAS 7440-27-9
Molecular Formula Tb
Concentration 100 mg/L
MDL Number MFCD00011256
InChI Key GZCRRIHWUXGPOV-UHFFFAOYSA-N
SMILES [Tb]
IUPAC Name terbium
Solvent or Matrix Nitric Acid
Molecular Weight (g/mol) 158.93
PubChem CID 23958
Quantity 100 mL
ChEBI CHEBI:33376
Chemical Name or Material Terbium
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.