missing translation for 'onlineSavingsMsg'
Learn More

Terephthalic acid, 98+%

Product Code. 11484184
Change view
Click to view available options
Quantity:
10,000 g
250 g
500 g
Unit Size:
10000g
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11484184 500 g 500g
11493950 250 g 250g
11494184 10,000 g 10000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11484184 Supplier Thermo Scientific Alfa Aesar Supplier No. A12527.36
 Restricted Item
In Stock
Add to basket
Zum Warenkorb hinzufügen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 100-21-0 | C8H6O4 | 166.132 g/mol

Terephthalic acid is used in the metal organic frame works. Terephthalic acid is used primarily to make polyethylene terephthalate (PET) resins and fibers, polyester fibers, films, polyethylene terephthalate solid-state resins and polyethylene terephthalate engineering resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Terephthalic acid is used in the metal organic frame works. Terephthalic acid is used primarily to make polyethylene terephthalate (PET) resins and fibers, polyester fibers, films, polyethylene terephthalate solid-state resins and polyethylene terephthalate engineering resins.

Solubility
slightly soluble in water (0,017 g/L at 25°C)

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It should be stored and used in molten form.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 100-21-0
Molecular Formula C8H6O4
Molecular Weight (g/mol) 166.132
MDL Number MFCD00002558
InChI Key KKEYFWRCBNTPAC-UHFFFAOYSA-N
Synonym p-phthalic acid, 1,4-benzenedicarboxylic acid, benzene-1,4-dicarboxylic acid, p-dicarboxybenzene, p-benzenedicarboxylic acid, p-carboxybenzoic acid, acide terephtalique, para-phthalic acid, tephthol, 1,4-dicarboxybenzene
PubChem CID 7489
ChEBI CHEBI:15702
IUPAC Name terephthalic acid
SMILES C1=CC(=CC=C1C(=O)O)C(=O)O

Spezifikation

Melting Point 402°C (sublimation)
Density 1.51
Flash Point 260°C (500°F)
Odor Odorless
Quantity 500 g
Beilstein 1909333
Merck Index 14,9162
Solubility Information slightly soluble in water (0,017g/L at 25°C)
Formula Weight 166.13
Percent Purity ≥98%
Chemical Name or Material Terephthalic acid
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.