missing translation for 'onlineSavingsMsg'
Learn More

ALDH1L1 Antibody (2E7), R&D Systems™

Product Code. 18621548
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.03mL
0.10mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18621548 0.025 mL 0.03mL
18630327 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18621548 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP250033SS
  • ONLINE EXCLUSIVE PRICE
    208.72 €
    172.20 € / 0.03mL
    Save 36.52 €
    17% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

ALDH1L1 Monoclonal antibody specifically detects ALDH1L1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALDH1L1
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 2E7
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:5000 - 1:10000, Immunohistochemistry 1:1000, Immunocytochemistry/ Immunofluorescence 1:1000
Formulation 50% PBS, 50% glycerol
Gene Accession No. O75891
Gene Alias 10-formyltetrahydrofolate dehydrogenase, aldehyde dehydrogenase 1 family, member L1, aldehyde dehydrogenase family 1 member L1, Cytosolic 10-formyltetrahydrofolate dehydrogenase, DKFZp781N0997, EC 1.5.1.6,10-FTHFDH, FDH, formyltetrahydrofolate dehydrogenase, FTHFD10-fTHF
Host Species Mouse
Immunogen Amino acids 402-902 of human ALDH1L1. [UniProt# O75891]
Purification Method Immunogen affinity purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Endocrinology, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10840
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.