missing translation for 'onlineSavingsMsg'
Learn More

TerpiMix 3 2500 μg/mL in Isopropanol, Dr. Ehrenstorfer

Product Code. 30326898
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30326898 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30326898 Supplier Dr. Ehrenstorfer Supplier No. DREA50000654IP
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (-)-Fenchone∣(-)-Trans-Caryophyllene∣(cis + trans)-Nerolidol∣alpha-Bisabolol∣alpha-Humulene∣Caryophyllene Oxide∣Cedrol∣Delta-3-Carene∣delta-Valerolactam∣Farnesol (Mixture of Isomers)∣Geraniol∣Geranyl Acetate∣Guaiol∣Limonene∣Nootkatone∣Phytane∣Phytol (cis/trans mixture)∣Squalene∣Valencene
CAS 7787-20-4, 87-44-5, 7212-44-4, 515-69-5, 6753-98-6, 1139-30-6
Analyte or Component Names (-)-Fenchone∣(-)-Trans-Caryophyllene∣(cis + trans)-Nerolidol∣alpha-Bisabolol∣alpha-Humulene∣Caryophyllene Oxide∣Cedrol∣Delta-3-Carene∣delta-Valerolactam∣Farnesol (Mixture of Isomers)∣Geraniol∣Geranyl Acetate∣Guaiol∣Limonene∣Nootkatone∣Phytane∣Phytol (cis/trans mixture)∣Squalene∣Valencene
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Isopropanol
Quantity 1 mL
Concentration or Composition (by Analyte or Components) (-)-Fenchone 2500 μg/mL∣(-)-Trans-Caryophyllene 2500 μg/mL∣(cis + trans)-Nerolidol 2500 μg/mL∣alpha-Bisabolol 2500 μg/mL∣alpha-Humulene 2500 μg/mL∣Caryophyllene Oxide 2500 μg/mL∣Cedrol 2500 μg/mL∣Delta-3-Carene 2500 μg/mL∣delta-Valerolactam 2500 μg/mL∣Farnesol (Mixture of Isomers) 2500 μg/mL∣Geraniol 2500 μg/mL∣Geranyl Acetate 2500 μg/mL∣Guaiol 2500 μg/mL∣Limonene 2500 μg/mL∣Nootkatone 2500 μg/mL∣Phytane 2500 μg/mL∣Phytol (cis/trans mixture) 2500 μg/mL∣Squalene 2500 μg/mL∣Valencene 2500 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.