missing translation for 'onlineSavingsMsg'
Learn More

±-Terpineol 1000 μg/mL in n-Hexane, Dr. Ehrenstorfer

Product Code. 30319019
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30319019 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30319019 Supplier Dr. Ehrenstorfer Supplier No. DREGA09010346HE
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material alpha-Terpineol
CAS 98-55-5
Analyte or Component Names alpha-Terpineol
Shipping Condition Room Temperature
Molecular Formula C10 H18 O
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym 3-Cyclohexene-1-methanol, α,α,4-trimethyl-, p-Menth-1-en-8-ol (8CI), (±)-α-Terpineol, 1,1-Dimethyl-1-(4-methylcyclohex-3-enyl)methanol, 1-p-Menthen-8-ol, 2-(4-Methyl-3-cyclohexenyl)-2-propanol, 4-(2-Hydroxy-2-propyl)-1-methylcyclohexene, 8-Hydroxy-p-menth-1-ene, Mil 350, NSC 21449, NSC 403665, PC 593, Pine Oil 593, Terpineol 350, dl-α-Terpineol, α,α,4-Trimethyl-3-cyclohexene-1-methanol, α-Terpineol
InChI Formula InChI=1S/C10H18O/c1-8-4-6-9(7-5-8)10(2,3)11/h4,9,11H,5-7H2,1-3H3
SMILES CC1=CCC(CC1)C(C)(C)O
IUPAC Name 2-(4-methylcyclohex-3-en-1-yl)propan-2-ol
Solvent or Matrix n-Hexane
Molecular Weight (g/mol) 154.25
Quantity 1 mL
Formula Weight 154.1358 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.