missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 2-bromobutyrate, 98%

Product Code. 11363257
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11363257 5 g 5g
11373257 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11363257 Supplier Thermo Scientific Alfa Aesar Supplier No. L13799.06
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 24457-21-4 | C8H15BrO2 | 223.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24457-21-4
Molecular Formula C8H15BrO2
Molecular Weight (g/mol) 223.11
MDL Number MFCD00209634
InChI Key RIUJWUWLGXBICR-UHFFFAOYSA-N
Synonym tert-butyl 2-bromobutyrate, tert-butyl2-bromobutanoate, t-butyl 2-bromobutyrate, 2-bromobutyric acid tert-butyl ester, 2-bromobutanoic acid tert-butyl ester, butanoic acid, 2-bromo-, 1,1-dimethylethyl ester, tertbutyl 2-bromobutyrate, tert-butyl-2-bromobutyrat, acmc-1cb0d, tert-butyl-2-bromobutyrate
PubChem CID 13124118
IUPAC Name tert-butyl 2-bromobutanoate
SMILES CCC(C(=O)OC(C)(C)C)Br

Specifications

Density 1.2
Boiling Point 72°C to 76°C (12 mmHg)
Refractive Index 1.441
Quantity 5 g
UN Number UN1760
Beilstein 2040298
Formula Weight 223.12
Percent Purity 98%
Chemical Name or Material tert-Butyl 2-bromobutyrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.