missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 2-bromoisobutyrate, 97%

Product Code. 11328045
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11328045

Brand: Thermo Scientific Alfa Aesar L08216.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 23877-12-5 | C8H15BrO2 | 223.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23877-12-5
Molecular Formula C8H15BrO2
Molecular Weight (g/mol) 223.11
MDL Number MFCD00051908
InChI Key IGVNJALYNQVQIT-UHFFFAOYSA-N
PubChem CID 90290
IUPAC Name tert-butyl 2-bromo-2-methylpropanoate
SMILES CC(C)(C)OC(=O)C(C)(C)Br

Specifications

Density 1.18
Boiling Point 35°C to 37°C (1 mmHg)
Refractive Index 1.436
Quantity 50 g
Beilstein 1905654
Synonym tert-butyl 2-bromoisobutyrate, t-butyl 2-bromo isobutyrate, 2-bromoisobutyric acid tert-butylester, tert-butyl 2-bromo-2-methylpropionate, tert-butyl alpha-bromoisobutyrate, t-butyl 2-bromoisobutylate, t-butyl 2-bromoisobutyrate, 2-bromoisobutyric acid tert-butyl ester, propanoic acid, 2-bromo-2-methyl-, 1,1-dimethylethyl ester, t-butyl2-bromoisobutyrate
Formula Weight 223.12
Percent Purity 97%
Chemical Name or Material tert-Butyl 2-bromoisobutyrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.