missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate, 97%, Thermo Scientific™

Product Code. 10065682
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10065682 1 g 1g
11396885 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10065682 Supplier Thermo Scientific Maybridge Supplier No. MO07066DA
 Restricted Item
Estimated Shipment: 15-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 185099-67-6
Molecular Formula C12H19NO3
Molecular Weight (g/mol) 225.288
InChI Key MENILFUADYEXNU-UHFFFAOYSA-N
Synonym tert-butyl 3-oxo-8-azabicyclo 3.2.1 octane-8-carboxylate, n-boc-nortropinone, n-boc-4-nortropinone, n-tert-butoxycarbonyl nortropinone, 8-boc-8-azabicyclo 3.2.1 octan-3-one, 3-oxo-8-aza-bicyclo 3.2.1 octane-8-carboxylic acid tert-butyl ester, t-butyl 3-oxo-8-azabicyclo 3.2.1 octane-8-carboxylate, 3-oxo-8-azabicyclo 3.2.1 octane-8-carboxylic acid tert-butyl ester, n-boc-nororopinone, 8-tert-butoxycarbonyl-8-azabicyclo 3.2.1 octan-3-one;
PubChem CID 2794767
IUPAC Name tert-butyl 3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate
SMILES CC(C)(C)OC(=O)N1C2CCC1CC(=O)C2

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 225.29
Percent Purity 97%
Chemical Name or Material tert-Butyl 3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.