missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl (3R)-3-aminobutanoate, 95%

Product Code. 10464605 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass Bottle
Unit Size:
2.5g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10464605 2.5 g Glass Bottle 2.5g
10542232 500 mg Glass Bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10464605 Supplier Thermo Scientific Acros Supplier No. 337950025
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 158849-23-1 | C8H17NO2 | 159.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 158849-23-1
Molecular Formula C8H17NO2
Molecular Weight (g/mol) 159.23
MDL Number MFCD00798293
InChI Key BFFNZGWJTHWUMY-ZCFIWIBFSA-N
Synonym tert-butyl 3r-3-aminobutanoate, r-tert-butyl 3-aminobutanoate, tert-butyl-3r-3-aminobutanoate, tert-butyl r-3-aminobutyrate, tert-butyl r-3-aminobutanoate, 1,1-dimethylethyl 3r-3-aminobutanoate, r-3-aminobutyric acid tert-butyl ester, tert-butyl r-3-amino-3-methylpropionate, butanoic acid, 3-amino-, 1,1-dimethylethyl ester, 3r
PubChem CID 2733826
IUPAC Name tert-butyl (3R)-3-aminobutanoate
SMILES CC(CC(=O)OC(C)(C)C)N

Specifications

Boiling Point 50.0°C (0.7 mmHg)
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Refractive Index 1.4240 to 1.4260
Specific Rotation − 24.00
Specific Rotation Condition −24° (20°C c=1,CHCl3)
Quantity 2.5 g
Formula Weight 159.23
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material tert-Butyl (3R)-3-aminobutanoate
Show More Show Less
compliance-icons
Product Identifier
  • tert-Butyl (3R)-3-aminobutanoate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.