missing translation for 'onlineSavingsMsg'
Learn More

tert-butyl 4-(4-bromobenzyloxy)piperidine-1-carboxylate, 97%, Thermo Scientific™

Product Code. 10317744
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10317744 1 g Amber Glass Bottle 1g
11376855 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10317744 Supplier Thermo Scientific Maybridge Supplier No. CC67910DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 930111-10-7
Molecular Formula C17H24BrNO3
Molecular Weight (g/mol) 370.287
MDL Number MFCD09879975
InChI Key HGSFDBSIEHXEQD-UHFFFAOYSA-N
Synonym tert-butyl 4-4-bromobenzyloxy piperidine-1-carboxylate, tert-butyl 4-4-bromobenzyl oxy piperidine-1-carboxylate, tert-butyl 4-4-bromophenyl methoxy piperidine-1-carboxylate, 4-4-bromobenzyloxy piperidine
PubChem CID 24229758
IUPAC Name tert-butyl 4-[(4-bromophenyl)methoxy]piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)OCC2=CC=C(C=C2)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 370.28
Percent Purity 97%
Chemical Name or Material Tert-butyl 4-(4-bromobenzyloxy)piperidine-1-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.