missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-[4-(hydroxymethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate, ≥97%, Thermo Scientific™

Product Code. 10289722
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10289722 1 g 1g
10729773 250 mg 250mg
10602485 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10289722 Supplier Thermo Scientific Maybridge Supplier No. CC30009DA
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 158985-37-6
Molecular Formula C16H24N2O3
Molecular Weight (g/mol) 292.379
MDL Number MFCD06411543
InChI Key PXKYXTXYWJKTSQ-UHFFFAOYSA-N
Synonym tert-butyl 4-4-hydroxymethyl phenyl piperazine-1-carboxylate, 4-4-n-boc-piperazinyl benzyl alcohol, tert-butyl 4-4-hydroxymethyl phenyl tetrahydro-1 2h-pyrazinecarboxylate, 4-4-n-boc-piperazinyl benzylalcohol, 4-4-tert-butoxycarbonyl piperazine benzyl alcohol, pubchem11318, buttpark 98\06-99, 4-1-boc-piperazin-4-yl phenylmethanol, 4-4-tert-butoxycarbonyl piperazin-1-yl benzyl alcohol
PubChem CID 2795510
IUPAC Name tert-butyl 4-[4-(hydroxymethyl)phenyl]piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=C(C=C2)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 292.38
Percent Purity 97%
Chemical Name or Material tert-Butyl 4-[4-(hydroxymethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.