missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-(bromomethyl)benzoate, 95%

Product Code. 15467537
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15467537 10 g 10g
15457537 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15467537 Supplier Thermo Scientific Alfa Aesar Supplier No. H35540.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108052-76-2 | C12H15BrO2 | 271.154 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108052-76-2
Molecular Formula C12H15BrO2
Molecular Weight (g/mol) 271.154
MDL Number MFCD04973450
InChI Key GSIBTIUXYYFCPU-UHFFFAOYSA-N
Synonym tert-butyl 4-bromomethyl benzoate, 4-bromomethylbenzoic acid tert-butyl ester, 4-bromomethyl-benzoic acid tert-butyl ester, 4-bromomethyl benzoic acid tert-butyl ester, tert-butyl p-bromomethyl benzoate, 4-bromo methyl benzoic acid mono tert-butyl ester, tert-butyl-4-bromomethylbenzoate, 1,1-dimethylethyl 4-bromomethyl benzoate, benzoic acid, 4-bromomethyl-, 1,1-dimethylethyl ester, 4-bromomethyl-benzoic acid, 1,1-dimethylethyl ester
PubChem CID 11414578
IUPAC Name tert-butyl 4-(bromomethyl)benzoate
SMILES CC(C)(C)OC(=O)C1=CC=C(C=C1)CBr

Specifications

Quantity 10 g
Formula Weight 271.15
Percent Purity 95%
Chemical Name or Material tert-Butyl 4-(bromomethyl)benzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.