missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl methyl sulfoxide, tech 85%, Thermo Scientific™

Product Code. 11327536
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11327536 1 g 1g
11337536 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11327536 Supplier Thermo Scientific Alfa Aesar Supplier No. L11081.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14094-11-2
Molecular Formula C5H12OS
Molecular Weight (g/mol) 120.21
MDL Number MFCD00041528
InChI Key YPRPSRLQZMDWFO-UHFFFAOYSA-N
Synonym tert-butyl methyl sulfoxide, tert-butyl methyl sulfoxide,-, 2-methyl-2-methylsulfinyl propane, unii-l3hjh0t7af, unii-08osz570vu, 2-methanesulfinyl-2-methylpropane, tert-butyl methylsulfoxide, l3hjh0t7af, tert-butyl methyl sulphoxide, r-methyl tert-butyl sulfoxide
PubChem CID 139684
IUPAC Name 2-methyl-2-methylsulfinylpropane
SMILES CC(C)(C)S(=O)C

Specifications

Boiling Point 64°C to 66°C (10 mmHg)
Refractive Index 1.473
Quantity 1 g
Beilstein 1815804
Formula Weight 120.21
Percent Purity 85%
Grade Technical
Chemical Name or Material tert-Butyl methyl sulfoxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.