missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl peroxybenzoate, 98%

Product Code. 11443617
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11443617 100 g 100g
11463950 25 g 25g
11453617 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11443617 Supplier Thermo Scientific Alfa Aesar Supplier No. A17373.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 614-45-9 | C11H14O3 | 194.23 g/mol

tert-Butyl peroxybenzoate was employed as polymerization and cross-linking catalyst. It was also was employed as initiator during  grafting of 2,2,6,6-tetramethyl-1-piperidinyloxy (TEMPO)-4-oxyacetamido-(3 propyltriethoxysilane) to poly(ethylene co-octene and in preparation of conformal poly(cyclohexyl methacrylate) thin films via initiated chemical vapor deposition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl peroxybenzoate was employed as polymerization and cross-linking catalyst. It was also was employed as initiator during  grafting of 2,2,6,6-tetramethyl-1-piperidinyloxy (TEMPO)-4-oxyacetamido-(3 propyltriethoxysilane) to poly(ethylene co-octene and in preparation of conformal poly(cyclohexyl methacrylate) thin films via initiated chemical vapor deposition.

Solubility
Soluble in ether, alcohol, ester, and ketones. Insoluble in water. 

Notes
 Stored and transported as a mixture with inert solids and as a solvent slurry.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 614-45-9
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00008802
InChI Key GJBRNHKUVLOCEB-UHFFFAOYSA-N
Synonym tert-butyl peroxybenzoate, tert-butyl perbenzoate, t-butyl perbenzoate, chaloxyd tbpb, perbutyl z, esperox 10, novox, trigonox c, tert-butyl peroxy benzoate, terc.butylperbenzoan
PubChem CID 11966
IUPAC Name tert-butyl benzenecarboperoxoate
SMILES CC(C)(C)OOC(=O)C1=CC=CC=C1

Specifikationer

Melting Point 6°C to 8°C
Density 1.042
Boiling Point 75°C to 76°C (0.2 mmHg)
Flash Point 93°C (199°F)
Odor Sweet
Refractive Index 1.499
Quantity 100 g
UN Number UN3103
Beilstein 1342734
Solubility Information Soluble in ether,alcohol,ester,and ketones. Insoluble in water.
Formula Weight 194.23
Percent Purity 98%
Chemical Name or Material tert-Butyl peroxybenzoate
Visa mer Visa mindre
compliance-icons compliance-icons compliance-icons
Produktbeteckning
  • tert-Butyl peroxybenzoate
Signalord
  • Danger
Riskkategorier
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Organic peroxide Type C & D
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Faroangivelser
  • H242-Heating may cause a fire.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H332-Harmful if inhaled.
  • H400-Very toxic to aquatic life.
Skyddsangivelse
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P411-Store at temperatures not exceeding °C/°F.
Kompletterande information
  • MIXTURE LIST-Contain: tert-Butyl peroxybenzoate

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.