missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl peroxybenzoate, 98%

Product Code. p-7023624
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11463950

Brand: Thermo Scientific Alfa Aesar A17373.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 614-45-9 | C11H14O3 | 194.23 g/mol

tert-Butyl peroxybenzoate was employed as polymerization and cross-linking catalyst. It was also was employed as initiator during  grafting of 2,2,6,6-tetramethyl-1-piperidinyloxy (TEMPO)-4-oxyacetamido-(3 propyltriethoxysilane) to poly(ethylene co-octene and in preparation of conformal poly(cyclohexyl methacrylate) thin films via initiated chemical vapor deposition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl peroxybenzoate was employed as polymerization and cross-linking catalyst. It was also was employed as initiator during  grafting of 2,2,6,6-tetramethyl-1-piperidinyloxy (TEMPO)-4-oxyacetamido-(3 propyltriethoxysilane) to poly(ethylene co-octene and in preparation of conformal poly(cyclohexyl methacrylate) thin films via initiated chemical vapor deposition.

Solubility
Soluble in ether, alcohol, ester, and ketones. Insoluble in water. 

Notes
 Stored and transported as a mixture with inert solids and as a solvent slurry.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 614-45-9
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00008802
InChI Key GJBRNHKUVLOCEB-UHFFFAOYSA-N
Synonym tert-butyl peroxybenzoate, tert-butyl perbenzoate, t-butyl perbenzoate, chaloxyd tbpb, perbutyl z, esperox 10, novox, trigonox c, tert-butyl peroxy benzoate, terc.butylperbenzoan
PubChem CID 11966
IUPAC Name tert-butyl benzenecarboperoxoate
SMILES CC(C)(C)OOC(=O)C1=CC=CC=C1

Specifications

Melting Point 6°C to 8°C
Density 1.042
Boiling Point 75°C to 76°C (0.2 mmHg)
Flash Point 93°C (199°F)
Odor Sweet
Refractive Index 1.499
Quantity 25 g
UN Number UN3103
Beilstein 1342734
Solubility Information Soluble in ether,alcohol,ester,and ketones. Insoluble in water.
Formula Weight 194.23
Percent Purity 98%
Chemical Name or Material tert-Butyl peroxybenzoate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • tert-Butyl peroxybenzoate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Organic peroxide Type C & D
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H242-Heating may cause a fire.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H332-Harmful if inhaled.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P411-Store at temperatures not exceeding 25°C/77°F.
Supplemental information
  • MIXTURE LIST-Contains : tert-Butyl peroxybenzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.